Assay Details
| Gene Symbol: Parp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007415.2 | NP_031441.2 | 6 - 7 | 894 | 75 | |
| GenBank mRNA | AK028042.1 | - | 6 - 7 | 913 | 75 | |
| AK030889.1 | - | 6 - 7 | 897 | 75 | ||
| AK088679.1 | - | 6 - 7 | 898 | 75 | ||
| AK146677.1 | - | 6 - 7 | 915 | 75 | ||
| AK152360.1 | - | 6 - 7 | 915 | 75 | ||
| AK159433.1 | - | 6 - 7 | 946 | 75 | ||
| AK165158.1 | - | 6 - 7 | 914 | 75 | ||
| AK166104.1 | - | 6 - 7 | 946 | 75 | ||
| AK167699.1 | - | 6 - 7 | 886 | 75 | ||
| AK169326.1 | - | 6 - 7 | 915 | 75 | ||
| AK210311.1 | - | 3 - 4 | 237 | 75 | ||
| BC012041.1 | - | 6 - 7 | 894 | 2648390 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp1
Genomic Map