Assay Details
| Gene Symbol: Parp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007415.2 | NP_031441.2 | 9 - 10 | 1362 | 99 | |
| GenBank mRNA | AK028042.1 | - | 9 - 10 | 1381 | 99 | |
| AK088679.1 | - | 9 - 10 | 1366 | 99 | ||
| AK146677.1 | - | 9 - 10 | 1383 | 99 | ||
| AK152360.1 | - | 9 - 10 | 1383 | 99 | ||
| AK159433.1 | - | 9 - 10 | 1414 | 99 | ||
| AK165158.1 | - | 9 - 10 | 1382 | 99 | ||
| AK166104.1 | - | 9 - 10 | 1414 | 99 | ||
| AK167699.1 | - | 9 - 10 | 1354 | 99 | ||
| AK169326.1 | - | 9 - 10 | 1383 | 99 | ||
| BC012041.1 | - | 9 - 10 | 1362 | 2648390 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp1
Genomic Map