Assay Details
| Gene Symbol: Anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007585.3 | NP_031611.1 | 9 - 10 | 756 | 88 | |
| GenBank mRNA | AK088645.1 | - | 9 - 10 | 745 | 88 | |
| AK146096.1 | - | 9 - 10 | 744 | 88 | ||
| AK150586.1 | - | 9 - 10 | 744 | 88 | ||
| AK152561.1 | - | 9 - 10 | 744 | 88 | ||
| AK164507.1 | - | 9 - 10 | 749 | 88 | ||
| AK165888.1 | - | 9 - 10 | 744 | 88 | ||
| AK167894.1 | - | 9 - 10 | 743 | 88 | ||
| AK168083.1 | - | 9 - 10 | 744 | 88 | ||
| BC003327.1 | - | 9 - 10 | 756 | 3156634 | 88 | |
| BC004659.1 | - | 5 - 6 | 1652 | 3497991 | 88 | |
| BC005763.1 | - | 9 - 10 | 730 | 2655513 | 88 | |
| D10024.1 | - | 8 - 9 | 689 | 88 | ||
| M14044.1 | - | 8 - 9 | 689 | 88 | ||
More Information
Set Membership: |
|

Anxa2
Genomic Map