Assay Details
| Gene Symbol: Canx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110499.1 | NP_001103969.1 | 3 - 4 | 434 | 120 | |
| NM_001110500.1 | NP_001103970.1 | 3 - 4 | 402 | 120 | ||
| NM_007597.3 | NP_031623.1 | 3 - 4 | 438 | 120 | ||
| GenBank mRNA | AK084175.1 | - | 3 - 4 | 378 | 120 | |
| AK159298.1 | - | 3 - 4 | 422 | 120 | ||
| AK165508.1 | - | 3 - 4 | 337 | 120 | ||
| AK166649.1 | - | 3 - 4 | 415 | 120 | ||
| BC012408.1 | - | 3 - 4 | 356 | 3497769 | 120 | |
| BC040244.1 | - | 3 - 4 | 365 | 2582119 | 120 | |
| L18888.1 | - | 2 - 3 | 250 | 120 | ||
| L23865.1 | - | 2 - 3 | 190 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Canx
Genomic Map