Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007998.6 | NP_032024.2 | 1 - 2 | 167 | 90 | |
| GenBank mRNA | AK004718.2 | - | 1 - 2 | 168 | 90 | |
| AK018738.2 | - | 1 - 2 | 170 | 90 | ||
| AK083560.1 | - | 1 - 2 | 133 | 90 | ||
| AK084843.1 | - | 1 - 2 | 65 | 90 | ||
| AK150685.1 | - | 1 - 2 | 169 | 90 | ||
| AK159652.1 | - | 1 - 2 | 200 | 90 | ||
| BC006746.1 | - | 1 - 2 | 174 | 3256604 | 90 | |
| BC094548.1 | - | 1 - 2 | 141 | 90 | ||
| M61215.1 | - | 1 - 2 | 160 | 90 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK084843.1 transcripts.
Set Membership: |
|

Fech
Genomic Map