Assay Details
| Gene Symbol: Fech | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007998.6 | NP_032024.2 | 6 - 7 | 799 | 79 | |
| GenBank mRNA | AK004718.2 | - | 6 - 7 | 800 | 79 | |
| AK018738.2 | - | 6 - 7 | 802 | 79 | ||
| AK084843.1 | - | 6 - 7 | 697 | 79 | ||
| AK150685.1 | - | 6 - 7 | 801 | 79 | ||
| AK159652.1 | - | 6 - 7 | 832 | 79 | ||
| BC094548.1 | - | 6 - 7 | 772 | 79 | ||
| BC096770.1 | - | 5 - 6 | 633 | 79 | ||
| M59288.1 | - | 5 - 6 | 636 | 79 | ||
| M61215.1 | - | 6 - 7 | 786 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fech
Genomic Map