Assay Details
| Gene Symbol: Hsd17b4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008292.4 | NP_032318.2 | 13 - 14 | 1364 | 67 | |
| GenBank mRNA | AK004866.1 | - | 13 - 14 | 1364 | 67 | |
| AK088381.1 | - | 13 - 14 | 1339 | 67 | ||
| AK147182.1 | - | 13 - 14 | 1336 | 67 | ||
| AK152532.1 | - | 13 - 14 | 1259 | 67 | ||
| AK161655.1 | - | 13 - 14 | 1259 | 67 | ||
| AK166801.1 | - | 13 - 14 | 1277 | 67 | ||
| AK167060.1 | - | 13 - 14 | 1314 | 67 | ||
| AK169077.1 | - | 13 - 14 | 1266 | 67 | ||
| AK187517.1 | - | 1 - 2 | 255 | 67 | ||
| AK216937.1 | - | 1 - 2 | 255 | 67 | ||
| BC022175.1 | - | 13 - 14 | 1237 | 4209394 | 67 | |
| X89998.1 | - | 13 - 14 | 1225 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b4
Genomic Map