Assay Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008490.2 | NP_032516.2 | 1 - 2 | 183 | 70 | |
| GenBank mRNA | AI574112.1 | - | 1 - 2 | 183 | 70 | |
| AK004914.1 | - | 1 - 2 | 179 | 70 | ||
| AK149476.1 | - | 1 - 2 | 180 | 70 | ||
| BC028861.1 | - | 1 - 2 | 166 | 4212194 | 70 | |
| J05154.1 | - | 1 - 2 | 163 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lcat
Genomic Map