Assay Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008490.2 | NP_032516.2 | 3 - 4 | 449 | 67 | |
| GenBank mRNA | AK004914.1 | - | 3 - 4 | 445 | 67 | |
| AK149476.1 | - | 3 - 4 | 446 | 67 | ||
| BC028861.1 | - | 3 - 4 | 432 | 4212194 | 67 | |
| J05154.1 | - | 3 - 4 | 429 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lcat
Genomic Map