Assay Details
| Gene Symbol: Ly9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277968.1 | NP_001264897.1 | 5 - 6 | 1198 | 72 | |
| NM_008534.3 | NP_032560.2 | 6 - 7 | 1528 | 72 | ||
| NR_102726.1 | 6 - 7 | 1544 | 72 | |||
| GenBank mRNA | AB196804.1 | - | 6 - 7 | 1441 | 72 | |
| AB196805.1 | - | 6 - 7 | 1441 | 72 | ||
| AB196806.1 | - | 6 - 7 | 1441 | 72 | ||
| AB196807.1 | - | 6 - 7 | 1441 | 72 | ||
| AF244130.1 | - | 6 - 7 | 1528 | 72 | ||
| AF244131.1 | - | 6 - 7 | 1528 | 72 | ||
| AK042288.1 | - | 6 - 7 | 1467 | 72 | ||
| AK087345.1 | - | 6 - 7 | 1448 | 72 | ||
| AK088815.1 | - | 5 - 6 | 1147 | 72 | ||
| AK134106.1 | - | 6 - 7 | 1545 | 72 | ||
| AK150121.1 | - | 6 - 7 | 1454 | 72 | ||
| BC055380.1 | - | 6 - 7 | 1428 | 72 | ||
| BC095921.1 | - | 6 - 7 | 1451 | 4935844 | 72 | |
| M84412.1 | - | 6 - 7 | 1392 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ly9
Genomic Map