Assay Details
| Gene Symbol: Slc3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161413.1 | NP_001154885.1 | 3 - 4 | 972 | 99 | |
| NM_008577.4 | NP_032603.3 | 2 - 3 | 731 | 99 | ||
| GenBank mRNA | AB023408.1 | - | 2 - 3 | 686 | 99 | |
| AK090046.1 | - | 2 - 3 | 709 | 99 | ||
| AK156285.1 | - | 2 - 3 | 203 | 99 | ||
| AK161280.1 | - | 3 - 4 | 973 | 99 | ||
| AK165417.1 | - | 2 - 3 | 713 | 99 | ||
| AK165550.1 | - | 2 - 3 | 709 | 99 | ||
| AK183775.1 | - | 1 - 2 | 96 | 99 | ||
| AK215373.1 | - | 2 - 3 | 197 | 99 | ||
| BC065173.1 | - | 2 - 3 | 691 | 6415269 | 99 | |
| U25708.1 | - | 2 - 3 | 708 | 99 | ||
| X14309.1 | - | 2 - 3 | 673 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc3a2
Genomic Map