Assay Details
| Gene Symbol: Slc3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161413.1 | NP_001154885.1 | 5 - 6 | 1136 | 59 | |
| NM_008577.4 | NP_032603.3 | 4 - 5 | 895 | 59 | ||
| GenBank mRNA | AB023408.1 | - | 4 - 5 | 850 | 59 | |
| AK090046.1 | - | 4 - 5 | 873 | 59 | ||
| AK156285.1 | - | 4 - 5 | 367 | 59 | ||
| AK161280.1 | - | 5 - 6 | 1137 | 59 | ||
| AK165417.1 | - | 4 - 5 | 877 | 59 | ||
| AK165550.1 | - | 4 - 5 | 873 | 59 | ||
| AK183775.1 | - | 3 - 4 | 260 | 59 | ||
| AK215848.1 | - | 2 - 3 | 120 | 59 | ||
| BC065173.1 | - | 4 - 5 | 855 | 6415269 | 59 | |
| U25708.1 | - | 4 - 5 | 872 | 59 | ||
| X14309.1 | - | 4 - 5 | 837 | 59 | ||
More Information
Set Membership: |
|

Slc3a2
Genomic Map