Assay Details
| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198635.1 | NP_001185564.1 | 15 - 16 | 1981 | 73 | |
| NM_008635.2 | NP_032661.2 | 15 - 16 | 1957 | 73 | ||
| GenBank mRNA | AB098611.1 | - | 15 - 16 | 1863 | 73 | |
| AK014575.1 | - | 15 - 16 | 1874 | 73 | ||
| AK133267.1 | - | 15 - 16 | 2406 | 73 | ||
| BC052637.1 | - | 15 - 16 | 1917 | 6467153 | 73 | |
| Y15197.1 | - | 15 - 16 | 1863 | 73 | ||
More Information
Set Membership: |
|

Map7
Genomic Map