Assay Details
| Gene Symbol: Nat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008673.1 | NP_032699.1 | 1 | 859 | 88 | |
| GenBank mRNA | BC119181.1 | - | 1 | 664 | 8733930 | 88 |
| BC137747.1 | - | 1 | 664 | 8860766 | 88 | |
| U35885.1 | - | 1 | 859 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160500 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | U35885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | U35885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937463 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937456 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937455 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937490 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937459 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937494 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937454 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937457 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937453 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937470 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937464 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937496 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937461 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937472 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937478 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937468 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937475 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25937488 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160500 | - | - |
| Whole Mouse Genome G4122A | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | U35885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | BC119181.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | NM_008673.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P317505 | BC137747.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103626 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160500 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07278767 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07278757 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07278759 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06945518 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07278771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937509 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25937549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937462 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937479 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937461 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06945518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937451 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937492 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937463 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937452 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937471 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937475 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937494 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937454 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07197132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937466 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937472 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937458 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937449 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937470 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07431193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07431204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937473 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25937455 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10572087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10572086 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421758_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10572085 | - | - |
| Mouse Genome 430 2.0 Array | 1421758_at | - | - |
| Mouse Genome 430 2.0 Array | 1421758_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17960_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 17960_at | - | - |
| Affymetrix Murine Genome 6500A Array | Msa16720_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800004633.mm.1 | NM_008673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004633.mm.1 | NM_008673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004633.mm.1 | TC0800000693.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800004634.mm.1 | NM_008673.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004634.mm.1 | NM_008673.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800004634.mm.1 | TC0800000693.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421758_PM_at | - | - |
| Mouse Expression Set 430 | 1421758_at | - | - |
| Murine Genome U74v2 Set | 94723_at | - | - |
| Murine Genome U74v2 Set | 94723_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421758_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421758_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421758_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421758_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 726231 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2480348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002480348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1225773 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1225773 | - | - |
| mouse-6 v1.1 (Illumina) | scl33761.1.1_187-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2480348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33761.1.1_187-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2480348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2879329 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2879329 | - | - |

Nat1
Genomic Map