Assay Details
| Gene Symbol: Nmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008707.3 | NP_032733.1 | 3 - 4 | 394 | 70 | |
| GenBank mRNA | AF043326.1 | - | 3 - 4 | 406 | 70 | |
| AK146523.1 | - | 3 - 4 | 409 | 70 | ||
| BC016526.1 | - | 3 - 4 | 394 | 3589678 | 70 | |
| BC021635.1 | - | 3 - 4 | 408 | 3989540 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nmt1
Genomic Map