Assay Details
| Gene Symbol: Npas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008719.2 | NP_032745.2 | 11 - 12 | 1557 | 66 | |
| GenBank mRNA | BC109166.2 | - | 11 - 12 | 1557 | 40048145 | 66 |
| U77969.1 | - | 11 - 12 | 1631 | 66 | ||
More Information
Set Membership: |
|

Npas2
Genomic Map