Assay Details
| Gene Symbol: Obp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008754.2 | NP_032780.2 | 5 - 6 | 524 | 132 | |
| GenBank mRNA | AK017455.1 | - | 5 - 6 | 525 | 132 | |
| BC125514.1 | - | 5 - 6 | 511 | 40130029 | 132 | |
| BC125516.1 | - | 5 - 6 | 511 | 40130031 | 132 | |
More Information
Set Membership: |
|

Obp1a
Genomic Map