Assay Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008777.3 | NP_032803.2 | 7 - 8 | 1065 | 81 | |
| GenBank mRNA | AK145839.1 | - | 7 - 8 | 930 | 81 | |
| AK149519.1 | - | 7 - 8 | 931 | 81 | ||
| BC013458.1 | - | 7 - 8 | 923 | 4223155 | 81 | |
| X51942.1 | - | 7 - 8 | 890 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pah
Genomic Map