Assay Details
| Gene Symbol: Pah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008777.3 | NP_032803.2 | 9 - 10 | 1189 | 63 | |
| GenBank mRNA | AK145839.1 | - | 9 - 10 | 1054 | 63 | |
| BC013458.1 | - | 9 - 10 | 1047 | 4223155 | 63 | |
| X51942.1 | - | 9 - 10 | 1014 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pah
Genomic Map