Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 4 - 5 | 532 | 76 | |
| GenBank mRNA | L01991.1 | - | 641 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn3
Genomic Map