Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 13 - 14 | 1871 | 84 | |
| GenBank mRNA | AK140747.1 | - | 4 - 5 | 460 | 84 | |
| L01991.1 | - | 1980 | 84 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn3
Genomic Map