Assay Details
| Gene Symbol: Cntn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008779.2 | NP_032805.2 | 17 - 18 | 2487 | 78 | |
| GenBank mRNA | AK140747.1 | - | 8 - 9 | 1076 | 78 | |
| L01991.1 | - | 2596 | 78 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn3
Genomic Map