Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 13 - 14 | 1751 | 68 | |
| NM_008797.3 | NP_032823.2 | 12 - 13 | 1728 | 68 | ||
| GenBank mRNA | AK077665.1 | - | 4 - 5 | 556 | 68 | |
| AK128927.1 | - | 1678 | 68 | |||
| AK148328.1 | - | 13 - 14 | 1698 | 68 | ||
| AK170596.1 | - | 13 - 14 | 1711 | 68 | ||
| AK170871.1 | - | 13 - 14 | 1701 | 68 | ||
| AK172320.1 | - | 13 - 14 | 1679 | 68 | ||
| BC055030.1 | - | 13 - 14 | 1686 | 5053475 | 68 | |
| L09192.1 | - | 12 - 13 | 1704 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map