Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 15 - 16 | 2371 | 60 | |
| NM_008797.3 | NP_032823.2 | 14 - 15 | 2348 | 60 | ||
| GenBank mRNA | AK077665.1 | - | 6 - 7 | 1176 | 60 | |
| AK148328.1 | - | 15 - 16 | 2318 | 60 | ||
| AK170596.1 | - | 15 - 16 | 2331 | 60 | ||
| AK170871.1 | - | 15 - 16 | 2321 | 60 | ||
| AK172320.1 | - | 15 - 16 | 2299 | 60 | ||
| BC055030.1 | - | 15 - 16 | 2306 | 5053475 | 60 | |
| L09192.1 | - | 14 - 15 | 2324 | 60 | ||
| M97957.1 | - | 9 - 10 | 1496 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map