Assay Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162946.1 | NP_001156418.1 | 19 - 20 | 3295 | 61 | |
| NM_008797.3 | NP_032823.2 | 18 - 19 | 3272 | 61 | ||
| GenBank mRNA | AK077665.1 | - | 10 - 11 | 2100 | 61 | |
| AK128927.1 | - | 3069 | 61 | |||
| AK148328.1 | - | 19 - 20 | 3242 | 61 | ||
| AK170596.1 | - | 19 - 20 | 3255 | 61 | ||
| AK170871.1 | - | 19 - 20 | 3246 | 61 | ||
| AK172320.1 | - | 19 - 20 | 3223 | 61 | ||
| BC055030.1 | - | 19 - 20 | 3230 | 5053475 | 61 | |
| L09192.1 | - | 18 - 19 | 3248 | 61 | ||
| M97957.1 | - | 13 - 14 | 2420 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcx
Genomic Map