Assay Details
| Gene Symbol: Pde7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122759.2 | NP_001116231.1 | 2 - 3 | 526 | 116 | |
| NM_008802.3 | NP_032828.2 | 2 - 3 | 256 | 116 | ||
| GenBank mRNA | AK036517.1 | - | 2 - 3 | 214 | 116 | |
| AK154521.1 | - | 2 - 3 | 343 | 116 | ||
| AK168957.1 | - | 2 - 3 | 218 | 116 | ||
| AY007702.1 | - | 2 - 3 | 205 | 116 | ||
| BC062909.1 | - | 2 - 3 | 525 | 6815827 | 116 | |
| CV558025.1 | - | 2 - 3 | 256 | 116 | ||
| U68171.1 | - | 2 - 3 | 204 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde7a
Genomic Map