Assay Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045558.1 | NP_001039023.1 | 6 - 7 | 692 | 63 | |
| NM_008837.3 | NP_032863.2 | 6 - 7 | 751 | 63 | ||
| GenBank mRNA | AK050224.1 | - | 5 - 6 | 596 | 63 | |
| AK090155.1 | - | 684 | 63 | |||
| BC023088.1 | - | 6 - 7 | 658 | 63 | ||
| BC048788.1 | - | 6 - 7 | 637 | 6488048 | 63 | |
| CA468666.1 | - | 6 - 7 | 572 | 63 | ||
| CD363874.1 | - | 3 - 4 | 181 | 63 | ||
| Z46720.1 | - | 5 - 6 | 440 | 63 | ||
More Information
Set Membership: |
|

Pick1
Genomic Map