| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100015153.mm.1 |
NM_008976.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015153.mm.1 |
NM_008976.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015153.mm.1 |
TC0100001821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015154.mm.1 |
NM_008976.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015154.mm.1 |
NM_008976.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015154.mm.1 |
TC0100001821.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100015152.mm.1 |
NM_008976.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015152.mm.1 |
NM_008976.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100015152.mm.1 |
TC0100001821.mm.1 |
MTA TC |