Assay Details
| Gene Symbol: Ptpn22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008979.2 | NP_033005.1 | 11 - 12 | 1047 | 81 | |
| NR_104070.1 | 13 - 14 | 1072 | 81 | |||
| GenBank mRNA | AK153812.1 | - | 13 - 14 | 1073 | 81 | |
| BC055377.1 | - | 11 - 12 | 969 | 30077916 | 81 | |
| M90388.1 | - | 11 - 12 | 1032 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn22
Genomic Map