Assay Details
| Gene Symbol: Ptpn22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008979.2 | NP_033005.1 | 14 - 15 | 2018 | 71 | |
| NR_104070.1 | 16 - 17 | 2043 | 71 | |||
| GenBank mRNA | AK153812.1 | - | 16 - 17 | 2044 | 71 | |
| BC055377.1 | - | 14 - 15 | 1940 | 30077916 | 71 | |
| M90388.1 | - | 14 - 15 | 2003 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn22
Genomic Map