Assay Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163688.1 | NP_001157160.1 | 17 - 18 | 2177 | 88 | |
| NM_008980.2 | NP_033006.2 | 18 - 19 | 2285 | 88 | ||
| GenBank mRNA | AF332081.1 | - | 16 - 17 | 1890 | 88 | |
| AF332082.1 | - | 16 - 17 | 1890 | 88 | ||
| AJ431367.1 | - | 16 - 17 | 1890 | 88 | ||
| AK031690.1 | - | 17 - 18 | 2176 | 88 | ||
| AK165673.1 | - | 17 - 18 | 2175 | 88 | ||
| BC050930.1 | - | 17 - 18 | 2159 | 6811172 | 88 | |
| BC110683.1 | - | 17 - 18 | 2143 | 3965311 | 88 | |
| M36033.1 | - | 18 - 19 | 2023 | 88 | ||
| Z23055.1 | - | 1 - 2 | 48 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpra
Genomic Map