Assay Details
| Gene Symbol: Ptprj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135657.1 | NP_001129129.1 | 13 - 14 | 2723 | 77 | |
| NM_008982.5 | NP_033008.3 | 13 - 14 | 3007 | 77 | ||
| GenBank mRNA | AK147318.1 | - | 13 - 14 | 2928 | 77 | |
| AK147556.1 | - | 13 - 14 | 2928 | 77 | ||
| AY038891.1 | - | 13 - 14 | 2825 | 77 | ||
| D45212.1 | - | 13 - 14 | 2963 | 77 | ||
| DQ133576.1 | - | 13 - 14 | 2738 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptprj
Genomic Map