Assay Details
| Gene Symbol: Igdcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008988.2 | NP_033014.1 | 12 - 13 | 2201 | 68 | |
| GenBank mRNA | AF026465.1 | - | 12 - 13 | 2201 | 68 | |
| AK011916.1 | - | 12 - 13 | 2299 | 68 | ||
| AK051027.1 | - | 12 - 13 | 2262 | 68 | ||
| BC053057.1 | - | 12 - 13 | 2238 | 5698028 | 68 | |
| BC086481.1 | - | 12 - 13 | 2183 | 6505546 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igdcc3
Genomic Map