Assay Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088292.1 | - | 7 - 8 | 951 | 145 | |
| AK146423.1 | - | 7 - 8 | 902 | 145 | ||
| AK156370.1 | - | 7 - 8 | 930 | 145 | ||
| X60672.1 | - | 7 - 8 | 857 | 145 | ||
More Information
Set Membership: |
|

Rdx
Genomic Map