Assay Details
| Gene Symbol: Rela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009045.4 | NP_033071.1 | 2 - 3 | 382 | 64 | |
| GenBank mRNA | AK089489.1 | - | 2 - 3 | 127 | 64 | |
| AK144565.1 | - | 2 - 3 | 127 | 64 | ||
| AK154630.1 | - | 2 - 3 | 127 | 64 | ||
| AK155321.1 | - | 2 - 3 | 393 | 64 | ||
| AK155889.1 | - | 2 - 3 | 124 | 64 | ||
| AK157076.1 | - | 2 - 3 | 114 | 64 | ||
| AK158072.1 | - | 2 - 3 | 392 | 64 | ||
| BC003818.1 | - | 2 - 3 | 141 | 3489295 | 64 | |
| BC094053.1 | - | 2 - 3 | 382 | 3157927 | 64 | |
| DQ020177.1 | - | 1 - 2 | 43 | 64 | ||
| M61909.1 | - | 2 - 3 | 97 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rela
Genomic Map