Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 6 - 7 | 969 | 68 | |
| GenBank mRNA | AK036208.1 | - | 6 - 7 | 971 | 68 | |
| AK083571.1 | - | 6 - 7 | 940 | 68 | ||
| AK147790.1 | - | 6 - 7 | 973 | 68 | ||
| BC057018.1 | - | 6 - 7 | 948 | 5705637 | 68 | |
| X76088.1 | - | 6 - 7 | 876 | 68 | ||
More Information
Set Membership: |
|

Rfx1
Genomic Map