Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 10 - 11 | 1702 | 82 | |
| GenBank mRNA | AK037226.1 | - | 3 - 4 | 645 | 82 | |
| AK083571.1 | - | 10 - 11 | 1673 | 82 | ||
| AK147790.1 | - | 10 - 11 | 1707 | 82 | ||
| BC057018.1 | - | 10 - 11 | 1681 | 5705637 | 82 | |
| X76088.1 | - | 10 - 11 | 1609 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx1
Genomic Map