Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 12 - 13 | 1937 | 62 | |
| GenBank mRNA | AK037226.1 | - | 5 - 6 | 880 | 62 | |
| AK083571.1 | - | 12 - 13 | 1908 | 62 | ||
| AK147790.1 | - | 12 - 13 | 1942 | 62 | ||
| BC057018.1 | - | 12 - 13 | 1916 | 5705637 | 62 | |
| X76088.1 | - | 12 - 13 | 1844 | 62 | ||
More Information
Set Membership: |
|

Rfx1
Genomic Map