Assay Details
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 16 - 17 | 2417 | 77 | |
| GenBank mRNA | AK037226.1 | - | 9 - 10 | 1360 | 77 | |
| AK083571.1 | - | 16 - 17 | 2388 | 77 | ||
| AK147790.1 | - | 16 - 17 | 2422 | 77 | ||
| BC057018.1 | - | 16 - 17 | 2396 | 5705637 | 77 | |
| X76088.1 | - | 16 - 17 | 2324 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx1
Genomic Map