Assay Details
| Gene Symbol: Rfx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009056.2 | NP_033082.1 | 2 - 3 | 216 | 88 | |
| NM_027787.1 | NP_082063.1 | 2 - 3 | 216 | 88 | ||
| GenBank mRNA | AK044439.1 | - | 2 - 3 | 216 | 88 | |
| AK086559.1 | - | 2 - 3 | 148 | 88 | ||
| BC004654.1 | - | 2 - 3 | 233 | 3497910 | 88 | |
| X76089.1 | - | 2 - 3 | 171 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx2
Genomic Map