Assay Details
| Gene Symbol: Rfx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009056.2 | NP_033082.1 | 15 - 16 | 2055 | 113 | |
| NM_027787.1 | NP_082063.1 | 16 - 17 | 2130 | 113 | ||
| GenBank mRNA | AK044439.1 | - | 16 - 17 | 2130 | 113 | |
| BC004654.1 | - | 15 - 16 | 2072 | 3497910 | 113 | |
| X76089.1 | - | 15 - 16 | 2010 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rfx2
Genomic Map