Assay Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U95145.1 | - | 1 - 2 | 166 | 72 | |
| U95146.1 | - | 1 - 2 | 166 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the U95145.1, U95146.1 transcripts.
Set Membership: |
|

Akap1
Genomic Map