Assay Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042541.1 | NP_001036006.1 | 3 - 4 | 1938 | 89 | |
| NM_009648.2 | NP_033778.2 | 3 - 4 | 1901 | 89 | ||
| NR_104310.1 | 4 - 5 | 2017 | 89 | |||
| GenBank mRNA | AK076216.1 | - | 3 - 4 | 1940 | 89 | |
| AK132749.1 | - | 3 - 4 | 1912 | 89 | ||
| BC065135.1 | - | 3 - 4 | 1850 | 89 | ||
| U95145.1 | - | 4 - 5 | 2021 | 89 | ||
| U95146.1 | - | 3 - 4 | 1914 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Akap1
Genomic Map