Assay Details
| Gene Symbol: Car2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009801.4 | NP_033931.4 | 2 - 3 | 558 | 90 | |
| GenBank mRNA | AK002333.1 | - | 2 - 3 | 293 | 90 | |
| AK146015.1 | - | 2 - 3 | 294 | 90 | ||
| BC044806.1 | - | 2 - 3 | 278 | 90 | ||
| BC055291.1 | - | 2 - 3 | 278 | 6479187 | 90 | |
| BU946040.1 | - | 2 - 3 | 560 | 90 | ||
| K00811.1 | - | 2 - 3 | 259 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Car2
Genomic Map