Assay Details
| Gene Symbol: Car2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009801.4 | NP_033931.4 | 5 - 6 | 836 | 68 | |
| GenBank mRNA | AK002333.1 | - | 5 - 6 | 571 | 68 | |
| AK002498.1 | - | 4 - 5 | 305 | 68 | ||
| AK146015.1 | - | 5 - 6 | 572 | 68 | ||
| BC044806.1 | - | 5 - 6 | 557 | 68 | ||
| BC055291.1 | - | 5 - 6 | 556 | 6479187 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car2
Genomic Map