Assay Details
| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009952.2 | NP_034082.1 | 1 - 2 | 217 | 75 | |
| NM_133828.2 | NP_598589.2 | 1 - 2 | 217 | 75 | ||
| GenBank mRNA | AF448507.1 | - | 1 - 2 | 93 | 75 | |
| AF448508.1 | - | 745 | 75 | |||
| AK042595.1 | - | 1 - 2 | 190 | 75 | ||
| AK044992.1 | - | 1 - 2 | 32 | 75 | ||
| AK044993.1 | - | 1 - 2 | 32 | 75 | ||
| AK079223.1 | - | 1 - 2 | 213 | 75 | ||
| BC021649.1 | - | 1 - 2 | 150 | 4017170 | 75 | |
| BY782293.1 | - | 1 - 2 | 174 | 75 | ||
| M95106.1 | - | 1 - 2 | 127 | 75 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF448507.1,AF448508.1,AK044992.1,AK044993.1,BC021649.1,M95106.1,NM_009952.2,NM_133828.2 transcripts.
Set Membership: |
|

Creb1
Genomic Map