Assay Details
| Gene Symbol: Creb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009952.2 | NP_034082.1 | 3 - 4 | 485 | 85 | |
| GenBank mRNA | AF448507.1 | - | 3 - 4 | 361 | 85 | |
| AF448508.1 | - | 1013 | 85 | |||
| M95106.1 | - | 3 - 4 | 395 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Creb1
Genomic Map