Assay Details
| Gene Symbol: Hipk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301304.1 | NP_001288233.1 | 9 - 10 | 2277 | 70 | |
| NM_010432.2 | NP_034562.2 | 9 - 10 | 2387 | 70 | ||
| NR_125731.1 | 10 - 11 | 2402 | 70 | |||
| GenBank mRNA | AF071070.1 | - | 9 - 10 | 2277 | 70 | |
| AF071071.1 | - | 10 - 11 | 2402 | 70 | ||
| AF077658.1 | - | 9 - 10 | 2291 | 70 | ||
| AF170304.1 | - | 9 - 10 | 2187 | 70 | ||
| AK122333.1 | - | 2 - 3 | 284 | 70 | ||
| BC145697.1 | - | 9 - 10 | 2140 | 40131000 | 70 | |
| BC145699.1 | - | 9 - 10 | 2140 | 40131002 | 70 | |
| DQ279086.1 | - | 3 - 4 | 471 | 70 | ||
| DQ279087.1 | - | 3 - 4 | 473 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hipk1
Genomic Map