Assay Details
| Gene Symbol: Hipk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301304.1 | NP_001288233.1 | 10 - 11 | 2411 | 75 | |
| NM_010432.2 | NP_034562.2 | 10 - 11 | 2521 | 75 | ||
| NR_125731.1 | 11 - 12 | 2536 | 75 | |||
| GenBank mRNA | AF071070.1 | - | 10 - 11 | 2411 | 75 | |
| AF071071.1 | - | 11 - 12 | 2536 | 75 | ||
| AF077658.1 | - | 10 - 11 | 2422 | 75 | ||
| AF170304.1 | - | 10 - 11 | 2321 | 75 | ||
| AK122333.1 | - | 3 - 4 | 418 | 75 | ||
| BC145697.1 | - | 10 - 11 | 2274 | 40131000 | 75 | |
| BC145699.1 | - | 10 - 11 | 2274 | 40131002 | 75 | |
| DQ279086.1 | - | 4 - 5 | 605 | 75 | ||
| DQ279087.1 | - | 4 - 5 | 607 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hipk1
Genomic Map