Assay Details
| Gene Symbol: Hipk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301304.1 | NP_001288233.1 | 12 - 13 | 2730 | 111 | |
| NM_001301306.1 | NP_001288235.1 | 11 - 12 | 2705 | 111 | ||
| NM_010432.2 | NP_034562.2 | 12 - 13 | 2840 | 111 | ||
| NR_125731.1 | 13 - 14 | 2855 | 111 | |||
| GenBank mRNA | AF071070.1 | - | 12 - 13 | 2730 | 111 | |
| AF071071.1 | - | 13 - 14 | 2855 | 111 | ||
| AF077658.1 | - | 12 - 13 | 2741 | 111 | ||
| AF170303.1 | - | 11 - 12 | 2505 | 111 | ||
| AF170304.1 | - | 12 - 13 | 2640 | 111 | ||
| AK122333.1 | - | 5 - 6 | 737 | 111 | ||
| BC145697.1 | - | 12 - 13 | 2593 | 40131000 | 111 | |
| BC145699.1 | - | 12 - 13 | 2593 | 40131002 | 111 | |
| DQ279087.1 | - | 6 - 7 | 926 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hipk1
Genomic Map